missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Phosphate Buffered Saline

Product Code. 12960119
Change view
Click to view available options
Fill Weight:
20 to 20.3 g
250 to 254 g
Format:
300 mL Sirop Bottle
Universal Bottle
Unit Size:
Each
Pack of 24
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Fill Weight Format Unit Size
12960119 20 to 20.3 g Universal Bottle Pack of 24
12637238 250 to 254 g 300 mL Sirop Bottle Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12960119 Supplier Thermo Scientific™ Supplier No. BO0971G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Create a pH-balanced environment to aid organism recovery using Phosphate Buffered Saline.

Phosphate Buffered Saline (Dulbecco A) is a preparation of a balanced salt solution without calcium and magnesium

TRUSTED_SUSTAINABILITY

Specifications

Color Colorless
Content And Storage 2° to 25°C
Format Universal Bottle
Manufacturing Origin United Kingdom
Product Type Microbiology Media
Fill Weight 20 to 20.3 g
Packaging Type 24 universal bottles with screw cap closures in a box
Quantity 24/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.