missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Phosphate-buffered saline (DPBS, 1X), Dulbecco's formula, without calcium, without magnesium
Click to view available options
Quantity:
500 mL
1 L
Unit Size:
1 L
500mL
Description
An anticoagulant and blood preservative. Permits the storage of whole blood at refrigerator temperatures for approximately 10 weeks.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Physical Form | Liquid |
| Quantity | 500 mL |
| Packaging | Plastic bottle |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ser du en mulighed for forbedring?Del en indholdskorrektion