missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Phosphate-buffered saline (DPBS, 1X), Dulbecco's formula, without calcium, without magnesium

Product Code. 15444695
Change view
Click to view available options
Quantity:
500 mL
1 L
Unit Size:
1 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15444695 1 L 1 L
15434695 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15444695 Supplier Thermo Scientific Chemicals Supplier No. J67802.K2
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

An anticoagulant and blood preservative. Permits the storage of whole blood at refrigerator temperatures for approximately 10 weeks.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Physical Form Liquid
Quantity 1 L
Packaging Plastic bottle

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.