missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Phosphate-buffered saline (PBS, 10X), pH 7.4

Product Code. 15424499
Change view
Click to view available options
Quantity:
1 L
2 L
4 L
Unit Size:
1 L
2 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15424499 2 L 2 L
15414499 1 L 1 L
15434499 4 L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15424499 Supplier Thermo Scientific Chemicals Supplier No. J62036.K3
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Non-toxic to most cells and is used in biological research.

Phosphate-buffered saline (PBS, 10X) is an isotonic, non-toxic to most cells and is used in biological research. It is associated with ethylenediaminetetraacetic acid (EDTA) and is used to disengage attached and clumped cells. It also helps to maintain a constant pH.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phosphate Buffered Saline (PBS)
Color Colorless
Concentration 10 X
pH 7.4
Physical Form Liquid
Quantity 2 L
What is the composition of Phosphate-buffered saline (PBS, 10X), pH 7.4 (Cat. No. J62036.K7, J62036-K7, J62036.K2, J62036-K2, J62036.K3, J62036-K3)?

Here is the composition of Phosphate-buffered saline (PBS, 10X), pH 7.4 (Cat. No. J62036.K7, J62036-K7, J62036.K2, J62036-K2, J62036.K3, J62036-K3):

- Sodium chloride: 1.37 M
- Potassium chloride (KCl): 27 mM
- Phosphate buffer: 101.4 mM
- Potassium phosphate monobasic: 17.6 mM

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.