missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Phosphate-buffered saline (PBS, 10X), pH 7.4

Product Code. p-7082298
missing translation for 'orderingAttributeHoverText'
Quantity:
1 L
2 L
4 L
missing translation for 'unitSize'
1 L
2 L
4 L
This item is not returnable. View return policy

Product Code. 15414499

missing translation for 'mfr': Thermo Scientific Chemicals J62036.K2

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Non-toxic to most cells and is used in biological research.

Phosphate-buffered saline (PBS, 10X) is an isotonic, non-toxic to most cells and is used in biological research. It is associated with ethylenediaminetetraacetic acid (EDTA) and is used to disengage attached and clumped cells. It also helps to maintain a constant pH.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phosphate Buffered Saline (PBS)
Color Colorless
Concentration 10 X
pH 7.4
Physical Form Liquid
Quantity 1 L

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.