Learn More
Thermo Scientific Chemicals Phosphate-buffered saline (PBS, 10X), pH 7.4, for Western blot
Description
Phosphate-buffered saline (PBS, 10X), is an isotonic, non-toxic to most cells and is used in biological research. It is associated with ethylenediaminetetraacetic acid (EDTA) and used to disengage attached and clumped cells. It also helps to maintain a constant pH.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Chemical Name or Material | Phosphate Buffered Saline (PBS) |
| Color | Colorless |
| Concentration | 10 X |
| For Use With (Application) | For Western blot washing. |
| pH | 7.4 |
| Physical Form | Liquid |
| Quantity | 2 L |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.