Learn More
Thermo Scientific Chemicals Phosphate-buffered saline (PBS, 10X), with Triton∣r X-100
Description
Phosphate-buffered saline is used in biological research. PBS buffer is ideal for maintaining a constant pH, and since it is isotonic to cells. PBS is frequently used as a wash and dilution buffer, and is also used to take a reference spectrum when measuring protein adsorption. And also used in immunoassay procedures.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Chemical Name or Material | Phosphate Buffered Saline (PBS) |
| Concentration | 10 X |
| Physical Form | Liquid |
| Quantity | 500 mL |
Safety and Handling
- Phosphate-buffered saline (PBS, 10X), with Triton X-100
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contain: Poly(oxy-1,2-ethanediyl), .alpha.-[4-(1,1,3,3-tetramethylbutyl)phenyl]- .omega.-hydroxy-
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Política de privacidad.