missing translation for 'onlineSavingsMsg'
Learn More

Phosphate solution for Ion Chromatography 1g/L H2O matrix, Fisher chemical™
BIOPHARMA

Product Code. 12912185 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 mL
250 mL
500 mL
Packaging:
Bottle
Unit Size:
100mL
250mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
12912185 100 mL Bottle 100mL
12922185 250 mL Bottle 250mL
12932185 500 mL Bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12912185 Supplier Fisher Chemical Supplier No. A 038 100
  • ONLINE EXCLUSIVE PRICE
    60.10 €
    51.16 € / 100mL
    Save 8.94 €
    14% Off
View all promotions
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Chemical Identifiers

CAS 7558-80-7
Molecular Weight (g/mol) 119.98
InChI Key AJPJDKMHJJGVTQ-UHFFFAOYSA-M
SMILES [Na+].OP(O)([O-])=O

Specifica

Packaging Bottle
Physical Form Liquid
Molecular Formula PO43-
Concentration 1 g/L
Type Single-component Standard Solution
IUPAC Name sodium dihydrogen phosphate
Solvent or Matrix Water
Quantity 100 mL
Chemical Name or Material Phosphate
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.