missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-Beclin 1 (Ser90) Recombinant Rabbit Monoclonal Antibody (1O6D0)

Rabbit Recombinant Monoclonal Antibody
176.00 € - 436.00 €
Specifications
| Antigen | Phospho-Beclin 1 (Ser90) |
|---|---|
| Clone | 1O6D0 |
| Concentration | 0.233 mg/mL |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
| Applications | ELISA, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30284551
|
Invitrogen™
MA558243 |
20 μL |
176.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30282703
![]() |
Invitrogen™
MA558244 |
100 μL |
436.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Phospho-Beclin 1 (Ser90) Recombinant Monoclonal Antibody for IHC (P), ELISA
Beclin 1 plays a central role in autophagy. It acts as a core subunit of the PI3K complex that mediates formation of phosphatidylinositol 3-phosphate. Different PI3K complex forms are believed to play a role in multiple membrane trafficking pathways - PI3KC3-C1 is involved in the initation of autophagosomes and PI3KC3-C2 is involved in the maturation of autophagosomes and endocytosis. Beclin 1 also plays a role in antiviral host defense.Specifications
| Phospho-Beclin 1 (Ser90) | |
| 0.233 mg/mL | |
| ELISA, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| O88597, Q14457, Q91XJ1 | |
| 114558, 56208, 8678 | |
| A synthetic phosphorylated peptide around S90 of human Beclin 1 (NP_003757.1). | |
| Antibody |
| 1O6D0 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| BECN1 | |
| 4921513J16Rik; 5430417M23Rik; APG6; Atg6; ATG6 autophagy related 6 homolog; Bcl-2-interacting protein beclin; beclin 1; beclin 1 (coiled-coil, moesin-like BCL2-interacting protein); beclin 1 (coiled-coil, myosin-like BCL2 interacting protein); beclin 1 (coiled-coil, myosin-like BCL2-interacting protein); beclin 1, autophagy related; beclin1; beclin-1; Beclin-1; beclin-1; Beclin-1-C 35 kDa; Beclin-1-C 37 kDa; becn1; Coiled-coil myosin-like BCL2-interacting protein; coiled-coil, myosin-like BCL2-interacting protein; GT197; hypothetical protein MGC73097; myosin-like BCL2 interacting protein; Protein GT197; testis secretory sperm-binding protein Li 215e; VPS30; zgc:73097 | |
| BECN1 | |
| Primary | |
| Protein A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title