missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-c-Cbl (Tyr774) Recombinant Rabbit Monoclonal Antibody (CblY774-R4C5), Invitrogen™
Shop All Thermo Scientific Products

Description
Cbl proteins are a family of ubiquitin protein ligases (E3s) that negatively regulate signaling by targeting activated tyrosine kinases for degradation. Cbl- c is the most recently cloned member of the Cbl proteins and is expressed only in epithelial cells (the other Cbl proteins are ubiquitously expressed). Cbl-c, like the other mammalian Cbl proteins, can ubiquitinate the activated EGFR and target it for degradation. Through interactions with proteins containing SRC homology-2 (SH2) and SH3 domains, CBL proteins modulate downstream cell signaling.
Specifications
Specifications
| Antigen | Phospho-c-Cbl (Tyr774) |
| Applications | Flow Cytometry, Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | CblY774-R4C5 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.10% BSA, 50% glycerol and 0.02% sodium azide |
| Gene | Cbl |
| Gene Accession No. | P22681, P22682 |
| Gene Alias | 4732447J05Rik; Cas-Br-M (murine) ecotropic retroviral transforming sequence; Casitas B-lineage lymphoma; casitas B-lineage lymphoma proto-oncogene; CBL; Cbl proto-oncogene; Cbl proto-oncogene, E3 ubiquitin protein ligase; CBL2; Cbl-2; CBL-3; CBLC; c-Cbl; E3 ubiquitin-protein ligase CBL; FRA11B; fragile site, folic acid type, rare, fra(11)(q23.3); NSLL; oncogene CBL2; proto-oncogene c-Cbl; RGD1561386; RING finger protein 55; RING-type E3 ubiquitin transferase CBL; RNF55; RNF57; signal transduction protein CBL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?