Learn More
Phospho-CD44 (Ser706) Polyclonal Antibody
GREENER_CHOICE
Phospho-CD44 (Ser706) Polyclonal Antibody
Brand:  Invitrogen PA5104810
Product Code. 16671255
  • 489.00 € / 100µL
Description
Phospho-CD44 (Ser706) Polyclonal Antibody for Western Blot, ICC/IF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Phospho-CD44 (Ser706)
Polyclonal
Unconjugated
CD44
AU023126; AW121933; AW146109; Bovine CD44; CD44; CD44 antigen; CD44 antigen-like protein; CD44 molecule; CD44 molecule (Indian blood group); CD44 protein; CD44 variant; CD44A; CD44-like protein; CDW44; cell surface glycoprotein CD44; Cell surface glycoprotein CD44 (hyaluronate binding protein); cell-surface glycoprotein; chondroitin sulfate proteoglycan 8; CSPG8; ECMRIII; ECMR-III; Epican; extracellular matrix receptor III; extracellular matrix receptor-III; GP90 lymphocyte homing/adhesion receptor; HCELL; hematopoietic cell E- and L-selectin ligand; heparan sulfate proteoglycan; HERMES; Hermes antigen; homing function and Indian blood group system; HUTCH-I; hyaluronan receptor; Hyaluronate receptor; IN; LHR; Ly-24; lymphocyte antigen 24; lymphocyte surface antigen precursor CD44; MC56; MDU2; MDU3; METAA; MIC4; PGP I; Pgp1; PGP-1; PGPI; PGP-I; Phagocytic glycoprotein 1; phagocytic glycoprotein I; putative Bos taurus CD44; RHAMM; sCD 44; sCD44; soluble CD 44; soluble CD44
Rabbit
Sequential Chromatography
RUO
12505, 25406, 960
-20°C
Liquid
Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P15379, P16070, P26051
CD44
A synthesized peptide derived from human CD44(Accession P16070), corresponding to amino acid residues around phosphorylated Ser706.
100 μL
Primary
Human, Mouse, Rat