missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-CDK6 (Tyr13) Polyclonal Antibody
GREENER_CHOICE

Product Code. 17749943
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17749943 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17749943 Supplier Invitrogen™ Supplier No. PA5118525

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of CDK6 only when phosphorylated at Tyr13.

CDK6 functions as a cell-cycle initiator protein. This protein is coexpressed and copurified with CyclinD1. They are critical regulators of cell cycle progression. The prototype member of this family, p34cdc2, and a related protein cdk2, function late in the cycle while cdk4 and cdk6 are critically involved in G1 to S progression. CDK6 is essential for cell proliferation within the dentate gyrus of the hippocampus and the cuventricular zone of the lateral ventricles. Mutations in the gene results in microcephaly 12, primary, autosomal recessive (MCPH12).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-CDK6 (Tyr13)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CDK6
Gene Accession No. Q00534, Q64261
Gene Alias 5830411I20; AI504062; CDK6; CDKN6; Cell division protein kinase 6; CR2 protein kinase; Crk2; cyclin dependent kinase 6; cyclin-dependent kinase 6; MCPH12; MGC59692; PLSTIRE; Serine/threonine-protein kinase PLSTIRE
Gene Symbols CDK6
Host Species Rabbit
Immunogen A synthesized peptide derived from human CDK6(Accession Q00534), corresponding to amino acid residues around phosphorylated Tyr13.
Purification Method sequential chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1021, 114483, 12571
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.