missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-EIF2S1 (Ser51) Recombinant Superclonal™ Antibody (10HCLC)
Rabbit Recombinant Superclonal Antibody
Brand: Invitrogen™ 710292
This item is not returnable.
View return policy
Description
Phospho-EIF2S1 (Ser51) Recombinant Polyclonal Antibody for Western Blot, ICC/IF, ChIP
EIF2a (Eukaryotic translation initiation factor 2A) is a heterotrimer composed of three subunits (alpha, beta, and gamma). This translation initiation factor drives binding of initiator methionyl-tRNA to the 40S ribosome in an AUG-dependent manner to form the 43S pre-initiation complex. The polypeptide can be phosphorylated by related protein kinases activated in response to stress. Phosphorylated EIF2A inhibits EIF2B activity and prevents guanine nucleotide exchange.
Specifications
| Phospho-EIF2S1 (Ser51) | |
| Recombinant Superclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide | |
| P05198 | |
| EIF2S1 | |
| Phosphopeptide corresponding to amino acids 48-57 of human EIF2a [pS51]. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ChIP Assay, Western Blot, Immunocytochemistry | |
| 10HCLC | |
| Unconjugated | |
| EIF2S1 | |
| 0910001O23Rik; 2410026C18Rik; 35kDa; EIF2; EIF-2; eIF2 alpha; eIF2 alpha subunit; EIF2A; eIF-2A; eIF2alpha; EIF-2alpha; eIF-2-alpha; EIF2S1; eif2s1b; Eukaryotic translation initiation factor 2 subunit 1; eukaryotic translation initiation factor 2 subunit 1b; eukaryotic translation initiation factor 2 subunit alpha; eukaryotic translation initiation factor 2, subunit 1 alpha; eukaryotic translation initiation factor 2, subunit 1 alpha b; eukaryotic translation initiation factor 2, subunit 1 alpha, 35kDa; eukaryotic translation initiation factor 2A; fb19d01; translation initiation factor subunit; wu:fb19d01 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 1965 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction