missing translation for 'onlineSavingsMsg'
Learn More

Phospho-GAB2 (Tyr643) Polyclonal Antibody, Invitrogen™

Product Code. 17769933
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17769933 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17769933 Supplier Invitrogen Supplier No. PA5118517

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Phospho-GAB2 (Tyr643) Polyclonal antibody specifically detects Phospho-GAB2 (Tyr643) in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-GAB2 (Tyr643)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation Dulbecco's PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene GAB2
Gene Accession No. Q9EQH1, Q9UQC2, Q9Z1S8
Gene Alias AI463667; D130058I17Rik; GAB2; Grb2 associated binder 2; GRB2 associated binding protein 2; GRB2-associated binder 2; GRB2-associated binding protein 2; GRB2-associated-binding protein 2; GRB3-3; Growth factor receptor bound protein 2-associated protein 2; KIAA0571; p97; PH domain-containing adaptor molecule p97; pp100
Host Species Rabbit
Immunogen Peptide sequence around phosphorylation site of tyrosine 643(V-D-Y(p)-V-Q) derived from Human Gab2 .
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14389, 84477, 9846
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.