missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-IRAK1 (Ser376) Polyclonal Antibody
GREENER_CHOICE

Product Code. 15544551
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15544551 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15544551 Supplier Invitrogen™ Supplier No. PA537611

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control for IHC is human brain tissue.

IL-1 receptor associated kinases (IRAKs) are serine/threonine kinases involved in signaling via Toll/IL-1 pathways. It is rapidly recruited by MYD88 to the receptor-signaling complex upon TLR activation. IRAK's association with MYD88 leads to IRAK1 phosphorylation by IRAK4 and subsequent autophosphorylation and kinase activation. Alternatively, IRAK1 can phosphorylate TIRAP to promote its ubiquitination and subsequent degradation. IRAK1 also phosphorylates the interferon regulatory factor 7 (IRF7) to induce its activation and translocation to the nucleus. This results in the transcriptional activiation of type I IFN genes, which drive the cell in an antiviral state.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-IRAK1 (Ser376)
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Irak1
Gene Accession No. P51617
Gene Alias AA408924; Il1rak; interleukin 1 receptor associated kinase 1; interleukin-1 receptor-associated kinase 1; interleukin-1 receptor-associated kinase-1; IRAK; Irak1; IRAK-1; IRAK1-S; mPLK; pelle; Pelle homolog; pelle-like protein kinase; Plpk; RGD1563841
Gene Symbols Irak1
Host Species Rabbit
Immunogen Peptide sequence around phosphorylation site of Serine 376(Q-S-S(p)-M-V) derived from Human IRAK1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3654
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.