missing translation for 'onlineSavingsMsg'
Learn More

Phospho-JDP2 (Thr148) Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Product Code. 16622445
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16622445 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16622445 Supplier Invitrogen Supplier No. PA5106001

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Phospho-JDP2 (Thr148) Polyclonal antibody specifically detects Phospho-JDP2 (Thr148) in Human, Mouse, Rat samples. It is validated for Immunofluorescence, Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-JDP2 (Thr148)
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4
Gene JDP2
Gene Accession No. Q78E65, Q8WYK2, P97875
Gene Alias c-Jun dimerization protein 2; Jdp2; Jdp-2; Jun dimerization protein 2; Jundm2; Jundp2; progesterone receptor co-activator; TIF
Host Species Rabbit
Immunogen A synthesized peptide derived from human JDP-2 around the phosphorylation site of Thr148.
Purification Method sequential chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81703, 116674, 122953
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.