missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-KV2.1 (KCNB1) (Ser805) Polyclonal Antibody, Invitrogen™
Description
Phospho-KV2.1 (KCNB1) (Ser805) Polyclonal antibody specifically detects Phospho-KV2.1 (KCNB1) (Ser805) in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | Phospho-KV2.1 (KCNB1) (Ser805) |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | Kcnb1 |
| Gene Accession No. | P15387, Q03717, Q14721 |
| Gene Alias | Delayed rectifier potassium channel 1; delayed rectifier potassium channel Kv2.1; DRK1; DRK1PC; KCB1; Kcnb1; Kcr1-1; Kv2.1; mShab; potassium channel Kv2.1; potassium channel, voltage gated Shab-related subfamily B, member 1; potassium voltage gated channel, Shab-related subfamily, member 1; potassium voltage-gated channel subfamily B member 1; potassium voltage-gated channel, Shab-related subfamily, member 1; Shab; voltage-gated potassium channel subunit Kv2.1 |
| Host Species | Rabbit |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?