Learn More
Invitrogen™ Phospho-MEK1 (Ser298) Recombinant Rabbit Monoclonal Antibody (24GB285)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557585
 View more versions of this product
Product Code. 30275168
  • 198.00 € / 20µL
Description
Phospho-MEK1 (Ser298) Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
MAP2K1 (MEK1) kinase is a dual-specificity protein kinase that functions in mitogen-activated protein kinase (MAPK) cascades, controlling cell growth and differentiation. MAP2K1 is activated by a wide variety of growth factors and cytokines and also by membrane-dependent depolarization and calcium influx. Mek1 is activated by phosphorylation of serine 218 and 222 residues by Raf1. It is known to be involved in the signaling during stress activate response, apoptosis and proliferative induction by cytokines. Mutations in the gene can lead to cardiofaciocutaneous syndrome.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Phospho-MEK1 (Ser298)
Recombinant Monoclonal
Unconjugated
Map2k1
CFC3; dual specificity mitogen activated protein kinase kinase 1; dual specificity mitogen-activated protein kinase kinase 1; EC 2.7.12.2; ERK activator kinase 1; kinase MEK1; MAP kinase kinase 1; MAP kinase kinase or Erk Kinase, Dual specificity mitogen-activated protein kinase kinase, involved in ras mediated vulval induction, LEThal LET-537 (42.8 kD) (mek-2); MAP kinase/Erk kinase 1; Map2k1; map2k1.L; MAPK/ERK kinase 1; MAPKK 1; MAPKK1; MEK 1; Mek1; MEK2; mek-2; MEKK1; mitogen activated protein kinase kinase 1; mitogen-activated protein kinase kinase 1; mitogen-activated protein kinase kinase 1 L homeolog; MKK (Thr292); MKK (Thr386); MKK1; MKK2; MP2K1; Prkmk1; PRKMK2; protein kinase, mitogen activated, kinase 1, p45; protein kinase, mitogen-activated, kinase 1 (MAP kinase kinase 1); XELAEV_18018447mg
Rabbit
Affinity chromatography
RUO
170851, 26395, 5604
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB285
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P31938, Q01986, Q02750
Map2k1
A synthesized peptide derived from human Phospho-MEK1 (Ser298) (aa 291-305).
20 μL
Primary
Human, Mouse, Rat