missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-MKP1/MKP2 (Ser296, Ser318) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA539795
This item is not returnable.
View return policy
Description
There are ten mitogen-activated protein kinase (MAPK) phosphatases (MKPs) that act as negative regulators of MAPK activity in mammalian cells. MKP2 (DUSP4) is a dual threonine/tyrosine phosphatase that specifically dephosphorylates and inactivates MAPKs (ERK, JNK, p38). It contains one Rhodanese domain and one tyrosine-protein phosphatase domain. MKP2 is expressed in a variety of tissues, and is localized in the nucleus. Transcription factor E2F-1 acts as a transcriptional regulator of MKP2. E2F-1/MKP2 pathway mediates apoptosis under oxidative stress and that MKP2 suppresses tumor formation. MKP2 is a novel transcription target of p53 in mediating apoptosis by 10-bp palindrome motif (CTGGCGCCAG) in the MKP2 promoter.
Specifications
| Phospho-MKP1/MKP2 (Ser296, Ser318) | |
| Polyclonal | |
| Unconjugated | |
| DUSP4 | |
| Mitogen-activated protein kinase phosphatase 2, MAP kinase phosphatase 2, MKP-2, Dual specificity protein phosphatase hVH2, DUSP-4, DUSP 4, MKPII, MKP II, Dual specificity protein phosphatase 1, MAP kinase phosphatase-1, MKP-1, Protein-tyrosine phosphatase CL100, Dual specificity protein phosphatase hVH1, CL100, MKP1, PTPN10, VH1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 114856, 1843, 1846, 19252, 319520, 60587 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P28562, Q13115, P28563, Q8BFV3, Q62767, Q64623 | |
| DUSP1, DUSP4 | |
| A synthetic phosphopeptide derived from human MKP-1/2 around the phosphorylation site of Ser296/318 (I-I-SP-P-N) | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction