missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-mTOR (Ser2448) Recombinant Rabbit Monoclonal Antibody (24GB330)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557588
This item is not returnable.
View return policy
Description
Phospho-mTOR (Ser2448) Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
FRAP1 (mTOR) is a serine/threonine kinase that plays a critical role in cellular growth and proliferation. Perturbations in the mTOR/PI3-kinase/AKT pathway are associated with numerous forms of cancer. FRAP1 is also the target of rapamycin and its analogues, which are currently used as immunosuppressants and cancer therapeutics. Mutations affecting the gene results in Smith-Kingsmore syndrome.
Specifications
| Phospho-mTOR (Ser2448) | |
| Recombinant Monoclonal | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P42345, P42346, Q9JLN9 | |
| MTOR | |
| A synthesized peptide derived from human Phospho-mTOR (Ser2448) (aa 2441-2455). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB330 | |
| Unconjugated | |
| MTOR | |
| 2610315D21Rik; AI327068; angiopoietin-like factor CDT6; FK506 binding protein 12-rapamycin associated protein 1; FK506 binding protein 12-rapamycin associated protein 2; FK506-binding protein 12-rapamycin complex-associated protein 1; FKBP12-rapamycin complex-associated protein; FKBP12-rapamycin complex-associated protein 1; FKBP-rapamycin associated protein; FKBP-rapamycin associated protein (FRAP); FKBP-rapamycin-associated protein FRAP; flat; FRAP; FRAP1; FRAP2; Mammalian target of rapamycin; mechanistic target of rapamycin; mechanistic target of rapamycin (serine/threonine kinase); mechanistic target of rapamycin kinase; Mtor; m-TOR; mTORC1; Raft1; Rapamycin and FKBP12 target 1; rapamycin and FKBP12 target-1 protein; rapamycin associated protein FRAP2; rapamycin target protein 1; RAPT1; serine/threonine-protein kinase mTOR; SKS | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 2475, 56717, 56718 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction