missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-Paxillin (Tyr31) Recombinant Rabbit Monoclonal Antibody (HL3129)

Phospho-Paxillin (Tyr31) Recombinant Rabbit Monoclonal Antibody (HL3129)
Brand: Invitrogen MA556834
This item is not returnable.
View return policy
Description
Phospho-Paxillin (Tyr31) Recombinant Monoclonal Antibody for WB, ICC/IF
Specifications
| Phospho-Paxillin (Tyr31) | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P49023, Q8VI36 | |
| PXN | |
| Carrier-protein conjugated synthetic peptide surrounding phospho Tyr31 of human Paxillin. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| HL3129 | |
| Unconjugated | |
| PXN | |
| adaptor protein; AW108311; AW123232; FLJ16691; hCG_1778014; I79_001229; myocardial ischemic preconditioning associated protein 7; Pax; PAX-1; PAXI; Paxillin; paxillin a; paxillin alpha; pxn; pxna; testicular tissue protein Li 134; TGFB1I1; transforming growth factor beta 1 induced transcript 1; wu:fw71f12 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 19303, 5829 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction