missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat Phospho-PLC-gamma 1 (Y783) Antibody, R&D Systems™

Rabbit Monoclonal Antibody
184.00 € - 398.00 €
Specifications
| Antigen | PLC-gamma 1 |
|---|---|
| Clone | 1016D |
| Dilution | Immunocytochemistry 3-25 ug/mL |
| Applications | Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18718829
![]() |
R&D Systems
MAB74542-SP |
25 μg |
184.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18741959
![]() |
R&D Systems
MAB74542 |
100 μg |
398.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PLC-gamma 1 Monoclonal specifically detects PLC-gamma 1 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry.Specifications
| PLC-gamma 1 | |
| Immunocytochemistry 3-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
| EC 3.1.4.11, gamma 1 (formerly subtype 148), monophosphatidylinositol phosphodiesterase, NCKAP3,1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase gamma-1, phosphatidylinositol phospholipase C, phosphoinositidase C, phosphoinositide phospholipase C, Phosphoinositide phospholipase C-gamma-1, phospholipase C, gamma 1, phospholipase C-148, Phospholipase C-gamma-1, Phospholipase C-II, PLC11-phosphatidylinositol-4,5-bisphosphate phosphodiesterase gamma 1, PLC148, PLC-148, PLCG1, PLCgamma 1, PLC-gamma-1, PLC-II1-phosphatidyl-D-myo-inositol-4,5-bisphosphate, PPLCA, triphosphoinositide phosphodiesterase | |
| PLCG1 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 1016D | |
| Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P19174 | |
| 5335 | |
| Phosphopeptide containing the human/mouse/rat PLC-gamma 1 Y783 site. Accession # P19174 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS | |
| Detects human, mouse, and rat PLC-gamma 1 when phosphorylated at Y783. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title