missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-PTS (Ser19) Recombinant Rabbit Monoclonal Antibody (24GB800)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557167
This item is not returnable.
View return policy
Description
Phospho-PTS (Ser19) Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
The tetrahydrobiopterin (BH4) cofactor is essential for hepatic hydroxylase, which is involved in phenylalanine degradation and catecholamine and serotonin biosynthesis. BH4 is also an essential and limiting cofactor for all types of nitric oxide synthase. BH4 deficiency results in hyperphenylalaninemia and monoamine neurotransmitter depletion and is most commonly due to autosomal recessive mutation in 6-pyruvoyltetrahydropterin synthase (PTPS), the second enzyme for BH4 biosynthesis. The active site of PTPS consists of the pterin-anchoring Glu A107 neighbored by two catalytic motifs: a Zn(II) binding site and an inter-subunit catalytic triad formed by Cys A42, Asp B88 and His B89. The active site of PTPS undergoes a Zn and Mg-dependent reaction that includes a triphosphate elimination, a stereospecific reduction and the oxidation of both side hydroxyl groups. The catalytic triad of PTPS is involved in the deprotonation of the side-chain carbons of substrates. In addition, Ser 19 of human PTPS may be a substrate for cGMP-dependent protein kinase type II phosphorylation in vivo, which is essential for normal activity of PTPS.
Specifications
| Phospho-PTS (Ser19) | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Pts | |
| 6-pyruvoyl tetrahydrobiopterin synthase; 6-pyruvoyltetrahydropterin synthase; 6-pyruvoyl-tetrahydropterin synthase; PTP synthase; PTPS; PTS | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 19286, 29498, 5805 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB800 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P27213, Q03393, Q9R1Z7 | |
| Pts | |
| A synthesized peptide derived from human Phospho-PTS (Ser19) (aa 12-26). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction