Learn More
Invitrogen™ Phospho-RAD17 (Ser656) Recombinant Rabbit Monoclonal Antibody (24GB795)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557600
 View more versions of this product
Product Code. 30275758
  • 564.00 € / 100µL
Description
Phospho-RAD17 (Ser656) Recombinant Monoclonal Antibody for Western Blot, Flow
Rad17 is essential for sustained cell growth, maintenance of chromosomal stability, and ATR-dependent checkpoint activation upon DNA damage. It has a weak ATPase activity required for binding to chromatin. Rad17 also participates in the recruitment of the RAD1-RAD9-HUS1 complex and RHNO1 onto chromatin, and in CHEK1 activation. May also serve as a sensor of DNA replication progression, and may be involved in homologous recombination.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Phospho-RAD17 (Ser656)
Recombinant Monoclonal
Unconjugated
Rad17
CCYC; cell cycle checkpoint protein (RAD17); cell cycle checkpoint protein RAD17; DNA damage checkpoint control protein RAD17; DNA repair exonuclease RAD17; FLJ41520; hRad17; MmRad24; R24L; RAD1 homolog; RAD17; RAD17 (Ser647); RAD17 checkpoint clamp loader component; RAD17 homolog; RAD17 homolog (S. pombe); Rad17 like protein; Rad17-like protein; Rad17p; RAD17SP; RAD24; RF-C activator 1 homolog; RF-C/activator 1 homolog; YOR368W
Rabbit
Affinity chromatography
RUO
19356, 5884
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB795
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O75943, Q6NXW6
Rad17
A synthesized peptide derived from human Phospho-Rad17 (Ser656) (aa 649-663).
100 μL
Primary
Human, Mouse