missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-RSK1/RSK2 (Ser221, Ser227) Polyclonal Antibody
GREENER_CHOICE

Product Code. 30535525
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30535525 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30535525 Supplier Invitrogen™ Supplier No. PA5149486

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Phospho-RSK1/RSK2 (Ser221, Ser227) Polyclonal Antibody for Western Blot, IHC (F)

RSK1 (Ribosomal S6 Kinase 1) and RSK2 (Ribosomal S6 Kinase 2) are genes that encode members of the RSK family of serine/threonine kinases, which are downstream effectors of the extracellular signal-regulated kinase (ERK) signaling pathway. These kinases play critical roles in regulating various cellular processes, including growth, proliferation, survival, and differentiation. RSK1 and RSK2 are activated by phosphorylation in response to extracellular signals such as growth factors, hormones, and mitogens. Once activated, they phosphorylate a range of substrates, including transcription factors, ribosomal proteins, and other kinases, thereby influencing gene expression and protein synthesis. Dysregulation of RSK1 and RSK2 activity has been implicated in several pathological conditions, including cancer, where they can promote tumor growth and survival, and neurodevelopmental disorders, such as Coffin-Lowry syndrome, which is associated with mutations in the RSK2 gene. Understanding the function and regulation of RSK1 and RSK2 is essential for developing targeted therapies aimed at modulating their activity in disease contexts. Research into these kinases provides valuable insights into the broader mechanisms of ERK signaling and its impact on cellular physiology and disease progression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-RSK1/RSK2 (Ser221, Ser227)
Applications Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Host Species Rabbit
Immunogen Phosphopeptide containing human RSK1 S221 site.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.