missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-STAT5 alpha (Tyr694) Polyclonal Antibody
GREENER_CHOICE

Product Code. 10231873
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10231873 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10231873 Supplier Invitrogen™ Supplier No. 716900
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Phospho-STAT5 alpha (Tyr694) Polyclonal Antibody for ELISA, ICC/IF, IHC, Western Blot

STAT5 alpha is a member of the STAT family of transcription factors. In response to cytokines and growth factors, STAT family members are phosphorylated by the receptor associated kinases, and then form homo- or heterodimers that translocate to the cell nucleus where they act as transcription activators. STAT5 alpha is activated by, and mediates the responses of many cell ligands, such as IL2, IL3, IL7 GM-CSF, erythropoietin, thrombopoietin, and different growth hormones. Activation of STAT5 alpha in myeloma and lymphoma associated with a TEL/JAK2 gene fusion is independent of cell stimulus and has been shown to be essential for the tumorigenesis. The mouse counterpart of STAT5 alpha is found to induce the expression of BCL2L1/BCL-X(L), which suggests the antiapoptotic function of this protein in cells. STAT5 alpha, along with STAT5 beta, were identified in mouse. The amino acid sequences of STAT5 alpha and STAT5 beta show 96% sequence similarity, and both proteins are co expressed in most tissues in virgin and lactating mice. However, differential accumulation of STAT5 alpha and STAT5 beta mRNA has been reported for both muscle and mammary tissue. STAT5 alpha is critically involved in a variety of physiological functions, including reproduction, lactation, immune function and somatic growth.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-STAT5 alpha (Tyr694)
Applications ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene STAT5A
Gene Accession No. P42229, P42230, P42231, Q62771, Q95115
Gene Alias AA959963; mammary gland factor; mammary gland factor STAT5A; mammary gland factor; transcription factor; Mgf; Mpf; signal transducer and activator of transcription 5; signal transducer and activator of transcription 5a; signal transducer and activator of transcription factor 5A; STAT; STAT5; STAT5A; STAT5A, Mammary Gland Factor; STAT5A/MGF; STAT5B; STATA5
Gene Symbols STAT5A
Host Species Rabbit
Immunogen Synthetic tyrosine-phosphorylated peptide encompassing the conserved C-terminal tyrosine phosphorylation site (Y694) of murine STAT5. This peptide is conserved in human, bovine, sheep, and rat STAT5 proteins.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20850, 24918, 282375, 443419, 6776
Target Species Human, Mouse, Rat, Bovine, Sheep
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.