missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Phospho-STAT6 Y641 Antibody, R&D Systems™

Rat Monoclonal Antibody
196.00 € - 378.00 €
Specifications
| Antigen | STAT6 |
|---|---|
| Clone | 943015 |
| Dilution | Immunocytochemistry 5-25 ug/mL |
| Applications | Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18794909
![]() |
R&D Systems
MAB37172-SP |
25 μg |
196.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18739138
![]() |
R&D Systems
MAB37172 |
100 μg |
378.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
STAT6 Monoclonal specifically detects STAT6 in Human samples. It is validated for Immunocytochemistry.Specifications
| STAT6 | |
| Immunocytochemistry 5-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| D12S1644, EC 2.4.1.227, EC 2.7.7.6, IL-4 Stat, IL-4-STAT, signal transducer and activator of transcription 6, signal transducer and activator of transcription 6, interleukin-4 induced, STAT, interleukin-4 induced, STAT, interleukin4-induced, STAT6B, STAT6C, transcription factor IL-4 STAT | |
| STAT6 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 943015 | |
| Immunocytochemistry | |
| Unconjugated | |
| Rat | |
| Human | |
| P42226 | |
| 6778 | |
| Phosphopeptide containing the human STAT6 Y641 site Accession # P42226 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human STAT6 when phosphorylated at Y641. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title