missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-STXBP1 (Ser313) Polyclonal Antibody
GREENER_CHOICE

Product Code. 16661635
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16661635 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16661635 Supplier Invitrogen™ Supplier No. PA5105191

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of MUNC-18a only when phosphorylated at Ser313.

This gene encodes a syntaxin-binding protein. The encoded protein appears to play a role in release of neurotransmitters via regulation of syntaxin, a transmembrane attachment protein receptor. Mutations in this gene have been associated with infantile epileptic encephalopathy-4. Alternatively spliced transcript variants have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-STXBP1 (Ser313)
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Stxbp1
Gene Accession No. O08599, P61764, P61765
Gene Alias AI317162; AI326233; ANC18HA; EIEE4; FLJ37475; hUNC18; MGC146482; minisatellite 10g detected by probe MMS10; MMS10-G; Ms10g; MUNC18-1; Munc-18a; munc-18c; neuronal SEC1; NSEC1; n-sec1; NSEC1A; p67; Protein unc-18 homolog 1; protein unc-18 homolog A; rbSec1; Rb-sec1; rbSec1A; rbSec1B; rop; RP11-56D16.3; Sec1; Stxbp1; Sxtbp1; syntaxin binding protein 1; syntaxin-binding protein 1; UNC18; UNC-18 homolog; unc-18-1; Unc18-1; UNC18A; unc-18A; Unc18h
Gene Symbols Stxbp1
Host Species Rabbit
Immunogen A synthesized peptide derived from human STXBP1(Accession P61764), corresponding to amino acid residues around phosphorylated Ser313.
Purification Method Sequential chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20910, 25558, 6812
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.