missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-Tensin 1 (Tyr1326) Polyclonal Antibody
GREENER_CHOICE

Product Code. 15516991
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15516991 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15516991 Supplier Invitrogen™ Supplier No. PA540300

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody is predicted to react with mouse based on sequence homology.

Tensin is involved in the maintenance of cellular structure by anchoring Actin filaments at the focal adhesion via F-Actin binding and capping activities. However, tensin also contains a Src homology 2 (SH2) domain and has the ability to be phosphorylated. Tensin is phosphorylated on tyrosine, serine, and threonine residues, suggesting that it might participate in signal transduction cascades. These diverse characteristics in a single molecule indicate that tensin may be an important link between the cytoskeleton and signal transduction pathways.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-Tensin 1 (Tyr1326)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TNS1
Gene Accession No. Q9HBL0
Gene Alias 1110018I21Rik; 1200014E20Rik; AI648117; DKFZp586K0617; E030018G17Rik; E030037J05Rik; matrix-remodelling associated 6; Matrix-remodelling-associated protein 6; MGC88584; MST091; MST122; MST127; MSTP091; MSTP122; MSTP127; MXRA6; PPP1R155; protein phosphatase 1, regulatory subunit 155; tensin 1; tensin-1; TNS; TNS1
Gene Symbols TNS1
Host Species Rabbit
Immunogen Synthesized peptide derived from human Tensin-1 around the phosphorylation site of Y1326.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21961, 7145
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.