missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-Tyrosine Antibody (SPM102), Novus Biologicals™
Mouse Monoclonal Antibody
277.00 € - 474.00 €
Specifications
| Antigen | Phospho-Tyrosine |
|---|---|
| Clone | SPM102 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 1-2 ug/mL, Flow Cytometry 1-2 ug/million cells, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1-2 ug/mL, Immunoprecipitation 1-2 ug/500 ug protein lysate, Immunohistochemistry-Paraffin 1-2 ug/mL |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18490642
|
Novus Biologicals
NBP2-32990-0.1mg |
0.1 mg |
474.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18414152
|
Novus Biologicals
NBP2-32990-0.02mg |
0.02 mg |
277.00 €
0.02mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Phospho-Tyrosine Monoclonal antibody specifically detects Phospho-Tyrosine in Nonspecies specific samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)Specifications
| Phospho-Tyrosine | |
| 0.2 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Neuroscience, Phospho Specific, Signal Transduction, Tyrosine Kinases | |
| 10 mM PBS with 0.05% BSA | |
| Phosphotyrosine conjugated to KLH | |
| Primary | |
| Store at 4C. |
| SPM102 | |
| Western Blot 1-2 ug/mL, Flow Cytometry 1-2 ug/million cells, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1-2 ug/mL, Immunoprecipitation 1-2 ug/500 ug protein lysate, Immunohistochemistry-Paraffin 1-2 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Non-species specific | |
| 2 amino 3(4 hydroxyphenyl) propanoic acid, 4 hydroxyphenylalanine, phosphotyrosine, pTyrosine, pY20, Tyrosine | |
| IgG2b κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title