missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-Zyxin (Ser143) Polyclonal Antibody

Description
Antibody detects endogenous levels of Zyxin only when phosphorylated at Ser143.
Zyxin is involved in focal adhesions, which are actin-rich structures that enable cells to adhere to the extracellular matrix and at which protein complexes involved in signal transduction assemble. Zyxin is a zinc-binding phosphoprotein that concentrates at focal adhesions and along the actin cytoskeleton. Zyxin has an N-terminal proline-rich domain and three LIM domains in its C-terminal half. The proline-rich domain may interact with SH3 domains of proteins involved in signal transduction pathways while the LIM domains are likely involved in protein-protein binding. Zyxin may function as a messenger in the signal transduction pathway that mediates adhesion-stimulated changes in gene expression and may modulate the cytoskeletal organization of actin bundles. Alternative splicing results in multiple transcript variants that encode the same isoform.
Specifications
Specifications
| Antigen | Phospho-Zyxin (Ser143) |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | ZYX |
| Gene Accession No. | Q15942, Q62523 |
| Gene Alias | 9530098H06Rik; ESP-2; HED-2; LOW QUALITY PROTEIN: zyxin; R75157; ZYX; Zyxin; zyxin-2 |
| Gene Symbols | ZYX |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?