missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospholipase C like 1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94627-0.02ml
This item is not returnable.
View return policy
Description
Phospholipase C like 1 Polyclonal antibody specifically detects Phospholipase C like 1 in Mouse, Rat samples. It is validated for Western Blot
Specifications
| Phospholipase C like 1 | |
| Polyclonal | |
| Western Blot 1:200-1:2000 | |
| but catalytically inactive protein, inactive phospholipase C-like protein 1, MGC126580, MGC138190, phospholipase C, epsilon, phospholipase C-deleted in lung carcinoma, phospholipase C-like 1, phospholipase C-related but catalytically inactive protein, PLCE, PLCL, PLC-L1, PRIP | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 110-350 of human PLCL1 (NP_006217.3). DCISFMQAGCELKKVRPNSRIYNRFFTLDTDLQALRWEPSKKDLEKAKLDISAIKEIRLGKNTETFRNNGLADQICEDCAFSILHGENYESLDLVANSADVANIWVSGLRYLVSRSKQPLDFMEGNQNTPRFMWLKTVFEAADVDGNGIMLEDTSVELIKQLNPTLKEAKIRLKFKEIQKSKEKLTTRVTEEEFCEAFCELCTRPEVYFLLVQISKNKEYLDANDLMLFLEAEQGVTHITE | |
| 0.02 mL | |
| Signal Transduction | |
| 5334 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction