missing translation for 'onlineSavingsMsg'
Learn More

Phosphorus standard solution, for AAS, 1 mg/ml P in water

Product Code. 10226350 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10226350 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10226350 Supplier Thermo Scientific Acros Supplier No. 196185000
  • ONLINE EXCLUSIVE PRICE
    72.00 €
    67.20 € / 500mL
    Save 4.80 €
    6% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7723-14-0 | P | 30.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For atomic adsorption spectroscopy
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phosphorus standard solution
Name Note 1 mg/ml P in water
Packaging Plastic bottle
CAS 7723-14-0, 7732-18-5
Assay Percent Range 98%
Physical Form Solution
Molecular Formula P
Concentration 997 to 1003 μg/mL P in water (20°C)
MDL Number MFCD00133771
Color Blue/Green
InChI Key OAICVXFJPJFONN-UHFFFAOYSA-N
SMILES [P]
IUPAC Name phosphorus
Molecular Weight (g/mol) 30.97
PubChem CID 5462309
Quantity 500 mL
ChEBI CHEBI:30207
Formula Weight 30.97
Grade AAS
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.