missing translation for 'onlineSavingsMsg'
Learn More

Phosphorus Triphenyl, MP Biomedicals

Product Code. 11461916 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11461916 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11461916 Supplier MP Biomedicals Supplier No. 0520998780
 Restricted Item
Estimated Shipment: 23-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 603-35-0
Molecular Formula C18H15P
MDL Number MFCD00003043 MFCD20489348
Quantity 100 g
Synonym triphenylphosphine, triphenyl phosphine, phosphine, triphenyl, triphenylphosphorus, triphenyl-phosphane, triphenylphosphide, phosphorustriphenyl, trifenylfosfin, trifenylfosfin czech, triphenylphosphine resin
InChI Key RIOQSEWOXXDEQQ-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
IUPAC Name triphenylphosphane
Molecular Weight (g/mol) 262.29
PubChem CID 11776
Formula Weight 262.3
Chemical Name or Material Phosphorus Triphenyl
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.