missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHPT1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
188.00 € - 470.00 €
Specifications
| Antigen | PHPT1 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18625102
|
Novus Biologicals
NBP2-94440-0.02ml |
0.02 mL |
188.00 €
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18674682
|
Novus Biologicals
NBP2-94440-0.1ml |
0.1 mL |
470.00 €
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PHPT1 Polyclonal antibody specifically detects PHPT1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| PHPT1 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Lipid and Metabolism | |
| PBS (pH 7.3), 50% glycerol | |
| 29085 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| bA216L13.10, DKFZp564M173, EC 3.1.3,14 kDa phosphohistidine phosphatase, EC 3.1.3.-, HSPC141, phosphohistidine phosphatase 11700008C22Rik, phosphohistidine phosphatase 14kDa, PHP14CGI-202, Protein janus-A homolog, RP11-216L13.10, sex-regulated protein janus-a | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-125 of human PHPT1 (NP_054891.2). MAVADLALIPDVDIDSDGVFKYVLIRVHSAPRSGAPAAESKEIVRGYKWAEYHADIYDKVSGDMQKQGCDCECLGGGRISHQSQDKKIHVYGYSMAYGPAQHAISTEKIKAKYPDYEVTWANDGY | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title