missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHPT1 Antibody (PHPT1/12053), Novus Biologicals™
Mouse Monoclonal Antibody
256.00 € - 484.00 €
Specifications
| Antigen | PHPT1 |
|---|---|
| Clone | PHPT1/12053 |
| Concentration | 0.2 mg/ml |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30305015
|
Novus Biologicals
NBP3-44602-100ug |
100 μg |
484.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30301160
|
Novus Biologicals
NBP3-44602-20ug |
20 μg |
256.00 €
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PHPT1 Monoclonal antibody specifically detects PHPT1 in Human samples. It is validated for ImmunohistochemistrySpecifications
| PHPT1 | |
| 0.2 mg/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| bA216L13.10, DKFZp564M173, EC 3.1.3,14 kDa phosphohistidine phosphatase, EC 3.1.3.-, HSPC141, phosphohistidine phosphatase 11700008C22Rik, phosphohistidine phosphatase 14kDa, PHP14CGI-202, Protein janus-A homolog, RP11-216L13.10, sex-regulated protein janus-a | |
| Recombinant human PHPT1 protein (Uniprot: Q9NRX4). Exact sequence is proprietary. | |
| Primary | |
| Store at 4⪚C. |
| PHPT1/12053 | |
| Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Lipid and Metabolism | |
| 10 mM PBS with 0.05% BSA | |
| 29085 | |
| IgG | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title