missing translation for 'onlineSavingsMsg'
Learn More

Phthalates Mixture 560 1000-10000 μg/mL in Hexane, Dr. Ehrenstorfer

Product Code. 30313412
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30313412 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30313412 Supplier Dr. Ehrenstorfer Supplier No. DREA50000560HE
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dibutyl phthalate∣Diethyl phthalate∣Diisononyl Phthalate (mixture of branched chain isomers)∣Di-n-octyl phthalate∣Phthalic acid, benzylbutyl ester∣Phthalic acid, bis-2-ethoxyethyl ester∣Phthalic acid, bis-2-ethylhexyl ester∣Phthalic acid, bis-4-methyl-2-pentyl ester∣Phthalic acid, bis-allyl ester∣Phthalic acid, bis-cyclohexyl ester∣Phthalic acid, bis-hexyl ester∣Phthalic acid, bis-iso-butyl ester∣Phthalic acid, bis-isodecyl ester∣Phthalic acid, bis-iso-pentyl ester∣Phthalic acid, bis-isopropylester∣Phthalic acid, bis-methyl ester∣Phthalic acid, bis-methylglycol ester∣Phthalic acid, bis-n-pentyl ester∣Phthalic acid, bis-phenyl ester∣Phthalic acid, bis-propyl ester∣Phthalic acid,bis-butoxyethyl ester∣Phthalic acid,bis-n-heptyl ester
CAS 84-74-2, 84-66-2, 68515-48-0, 117-84-0, 85-68-7, 605-54-9
Quantity 1 mL
Concentration or Composition (by Analyte or Components) Dibutyl phthalate 1000 μg/mL∣Diethyl phthalate 1000 μg/mL∣Di-n-octyl phthalate 1000 μg/mL∣Phthalic acid, benzylbutyl ester 1000 μg/mL∣Phthalic acid, bis-2-ethoxyethyl ester 1000 μg/mL∣Phthalic acid, bis-2-ethylhexyl ester 1000 μg/mL∣Phthalic acid, bis-4-methyl-2-pentyl ester 1000 μg/mL∣Phthalic acid, bis-allyl ester 1000 μg/mL∣Phthalic acid, bis-cyclohexyl ester 1000 μg/mL∣Phthalic acid, bis-hexyl ester 1000 μg/mL∣Phthalic acid, bis-iso-butyl ester 1000 μg/mL∣Phthalic acid, bis-iso-pentyl ester 1000 μg/mL∣Phthalic acid, bis-isopropylester 1000 μg/mL∣Phthalic acid, bis-methyl ester 1000 μg/mL∣Phthalic acid, bis-methylglycol ester 1000 μg/mL∣Phthalic acid, bis-n-pentyl ester 1000 μg/mL∣Phthalic acid, bis-phenyl ester 1000 μg/mL∣Phthalic acid, bis-propyl ester 1000 μg/mL∣Phthalic acid,bis-butoxyethyl ester 1000 μg/mL∣Phthalic acid,bis-n-heptyl ester 1000 μg/mL∣Diisononyl Phthalate (mixture of branched chain isomers) 10000 μg/mL∣Phthalic acid, bis-isodecyl ester 10000 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.