missing translation for 'onlineSavingsMsg'
Learn More

Phthalic acid, bis-n-octyl ester, Dr. Ehrenstorfer

Product Code. 30312145
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30312145 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30312145 Supplier Dr. Ehrenstorfer Supplier No. DREC16175000100MG
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Di-n-octyl phthalate
CAS 117-84-0
Molecular Formula C24 H38 O4
Quantity 100 mg
Synonym Phthalic acid, bis-n-octyl ester
InChI Formula InChI=1S/C24H38O4/c1-3-5-7-9-11-15-19-27-23(25)21-17-13-14-18-22(21)24(26)28-20-16-12-10-8-6-4-2/h13-14,17-18H,3-12,15-16,19-20H2,1-2H3
SMILES CCCCCCCCOC(=O)c1ccccc1C(=O)OCCCCCCCC
IUPAC Name dioctyl benzene-1,2-dicarboxylate
Molecular Weight (g/mol) 390.56
Formula Weight 390.277 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.