missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phthalic acid, mono-2-ethyl-5-oxohexyl ester 100 μg/mL in Methyl-tert-butyl ether, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Phthalic acid, mono-2-ethyl-5-oxohexyl ester |
| CAS | 40321-98-0 |
| Molecular Formula | C16 H20 O5 |
| Quantity | 1 mL |
| Synonym | rac Mono(2-ethyl-5-oxohexyl) Phthalate, Phthalic acid mono-2-ethyl-5-oxohexyl ester |
| InChI Formula | InChI=1S/C16H20O5/c1-3-12(9-8-11(2)17)10-21-16(20)14-7-5-4-6-13(14)15(18)19/h4-7,12H,3,8-10H2,1-2H3,(H,18,19) |
| SMILES | CCC(CCC(=O)C)COC(=O)c1ccccc1C(=O)O |
| IUPAC Name | 2-(2-ethyl-5-oxohexoxy)carbonylbenzoic acid |
| Molecular Weight (g/mol) | 292.33 |
| Formula Weight | 292.1311 g/mol |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?