missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PI15 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA556325
This item is not returnable.
View return policy
Description
Immunogen sequence: PQDCNPRCPM RCFGPMCTHY TQMVWATSNR IGCAIHTCQN MNVWGSVWRR AVYLVCNYAP KGNWIGEAPY KVGVPCSSCP PSYGGSCTDN LCFPGVTSNY LY Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.
This gene encodes a trypsin inhibitor. The protein shares similarity to insect venom allergens, mammalian testis-specific proteins and plant pathogenesis-related proteins. It is frequently expressed in human neuroblastoma and glioblastoma cell lines, and thus may play a role in the central nervous system. [provided by RefSeq].
Specifications
| PI15 | |
| Polyclonal | |
| Unconjugated | |
| PI15 | |
| 25 kDa trypsin inhibitor; CRISP8; CRISP-8; Cysteine-rich secretory protein 8; DKFZp686F0366; P24TI; P25TI; Peptidase inhibitor 15; PI15; PI-15; protease inhibitor 15; sugarCrisp | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51050 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O43692 | |
| PI15 | |
| Recombinant protein corresponding to Human PI15. Recombinant protein control fragment (Product #RP-89268). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction