missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PI3K p101 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13329-1-AP-150UL
This item is not returnable.
View return policy
Description
Receptor-regulated class I phosphoinositide 3-kinases.Specifications
| PI3K p101 | |
| Polyclonal | |
| Unconjugated | |
| PIK3R5 | |
| F730038I15Rik, FOAP 2, p101, P101 PI3K, PI3 kinase p101 subunit, PI3K p101, PIK3R5, Protein FOAP 2, PtdIns 3 kinase p101 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23533, 320207, 497931 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5SW28, Q8WYR1 | |
| Pik3r5 | |
| PI3K p101 Fusion Protein Ag4137 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction