missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human PI4K2B (PI4K2 beta), GST Tag Recombinant Protein
Human PI4K2B (PI4K2 beta), GST Tag Recombinant Protein
Brand: Invitrogen™ A32906
Additional Details : Weight : 0.00900kg
Description
PI4K2B lipid kinase is a member of the phosphatidylinositol 4-kinase family (PI4K) that phosphorylate phosphatidylinositol to generate phosphatidylinositol 4-phosphate which is an immediate precursor of several important signaling and scaffolding molecules. PI4K2B is primarily a cytosolic PI4K that is recruited to membranes where it stimulates phosphatidylinositol 4,5-bisphosphate syntheses. PI4K2B uses phosphatidylinositol as its primary substrate and has no activity on phosphatidylinositol monophosphates. PI4K2B is a restricted minor histocompatibility antigen in patients that had been successfully treated with donor lymphocyte infusions for relapsed chronic myeloid leukemia after allogeneic stem cell transplantation.- Validated for use with the Adapta™ assay
- Available in multiple pack sizes for convenient use
Specifications
NP_060793 | |
82.1 kDa | |
PI3 Kinase | |
Human | |
GST Tag | |
Dry Ice | |
Full Length, Recombinant, Active |
Kinase Assay | |
100 μg | |
Full Length | |
Recombinant | |
Storage Buffer: 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol. | |
Insect | |
Liquid |