missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human PI4K2B (PI4K2 beta), GST Tag Recombinant Protein
Human PI4K2B (PI4K2 beta), GST Tag Recombinant Protein
846.00 € - 7580.00 €
Specifications
Accession Number | NP_060793 |
---|---|
For Use With (Application) | Kinase Assay |
Molecular Weight (g/mol) | 82.1 kDa |
KinaseFamily | PI3 Kinase |
Protein Length | Full Length |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15960313
|
Invitrogen™
A31080 |
20 μg |
846.00 €
20µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17148805
|
Invitrogen™
A33391 |
1 mg |
7580.00 €
1mg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17178755
|
Invitrogen™
A32906 |
100 μg |
2435.00 €
100µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
PI4K2B lipid kinase is a member of the phosphatidylinositol 4-kinase family (PI4K) that phosphorylate phosphatidylinositol to generate phosphatidylinositol 4-phosphate which is an immediate precursor of several important signaling and scaffolding molecules. PI4K2B is primarily a cytosolic PI4K that is recruited to membranes where it stimulates phosphatidylinositol 4,5-bisphosphate syntheses. PI4K2B uses phosphatidylinositol as its primary substrate and has no activity on phosphatidylinositol monophosphates. PI4K2B is a restricted minor histocompatibility antigen in patients that had been successfully treated with donor lymphocyte infusions for relapsed chronic myeloid leukemia after allogeneic stem cell transplantation.- Validated for use with the Adapta™ assay
- Available in multiple pack sizes for convenient use
Specifications
NP_060793 | |
82.1 kDa | |
Full Length | |
Recombinant | |
Storage Buffer: 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol. | |
Full Length, Recombinant, Active |
Kinase Assay | |
PI3 Kinase | |
Human | |
GST Tag | |
Dry Ice | |
Liquid |