missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PI4K2B (PI4K2 beta) Recombinant Human Protein
This PI4K2B (PI4K2 beta) Recombinant Human Protein (full length) was expressed in insect cells.
Brand: Invitrogen™ A31080
Additional Details : Weight : 0.01000kg
Description
- This PI4K2B (PI4K2 beta) recombinant human protein (full length) was expressed in insect cells
- PI4K2B lipid kinase is a member of the phosphatidylinositol 4-kinase family (PI4K)
- PI4K2B is primarily a cytosolic PI4K that is recruited to membranes where it stimulates phosphatidylinositol 4,5-bisphosphate syntheses
- It uses phosphatidylinositol as its primary substrate and has no activity on phosphatidylinositol monophosphates
- PI4K2B is a restricted minor histocompatibility antigen in patients that had been successfully treated with donor lymphocyte infusions for relapsed chronic myeloid leukemia after allogeneic stem cell transplantation
- PI4K2B (PI4K2 beta) recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service
- Validated for use with the Adapta™ assay
- Available in multiple pack sizes for convenient use
Specifications
NP_060793 | |
20 µg | |
Full Length | |
GST Tag | |
Dry Ice | |
Kinases & Inhibitors | |
Liquid | |
Human |
82.1 kDa | |
PI3 Kinase | |
Yes | |
Storage Buffer: 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol. | |
Baculovirus | |
Full Length, Recombinant, Active | |
Kinase Assay |