missing translation for 'onlineSavingsMsg'
Learn More

PIANO (DHA) Standard

Product Code. 30328652
Change view
Click to view available options
Quantity:
1 mL
10 x 1 mL
Unit Size:
10mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30328652 10 x 1 mL 10mL
30319783 1 mL -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30328652 Supplier VHG Labs Supplier No. VHGPIANODHA10X1
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,2,4,5-Tetramethylbenzene∣1,2,4-Trimethylbenzene∣2,2,4-Trimethylpentane∣2,2-Dimethylbutane∣2,3,3 Trimethyl-1-butene∣2,3-Dimethyl-1-butene∣2,3-Dimethylpentane∣2,4-Dimethylpentane∣2-Methyl-1-Heptene∣2-Methylbutane∣2-Methylpentane∣3-Ethyltoluene∣3-Methylpentane∣Benzene∣Cyclohexane∣Decahydronaphthalene∣Ethylbenzene∣Ethylcyclohexane∣Isobutylbenzene∣Isopropylbenzene∣Methylcyclohexane∣n-Butylbenzene∣n-Butylcyclohexane∣n-Decane∣n-Decene∣n-Dodecane∣n-Dodecene∣n-Heptane∣n-Heptene∣n-Hexane∣n-Hexene∣n-Nonane∣n-Nonene∣n-Octane∣n-Octene∣n-Pentane∣n-Pentene∣n-Pentylbenzene∣n-Pentylcyclohexane∣n-Undecane∣n-Undecene∣Propylbenzene∣Propylcyclohexane∣p-Xylene (1,4-Dimethylbenzene)∣Toluene
CAS 95-93-2, 95-63-6, 540-84-1, 75-83-2, 594-56-9, 563-78-0
Analyte or Component Names 1,2,4,5-Tetramethylbenzene∣1,2,4-Trimethylbenzene∣2,2,4-Trimethylpentane∣2,2-Dimethylbutane∣2,3,3 Trimethyl-1-butene∣2,3-Dimethyl-1-butene∣2,3-Dimethylpentane∣2,4-Dimethylpentane∣2-Methyl-1-Heptene∣2-Methylbutane∣2-Methylpentane∣3-Ethyltoluene∣3-Methylpentane∣Benzene∣Cyclohexane∣Decahydronaphthalene∣Ethylbenzene∣Ethylcyclohexane∣Isobutylbenzene∣Isopropylbenzene∣Methylcyclohexane∣n-Butylbenzene∣n-Butylcyclohexane∣n-Decane∣n-Decene∣n-Dodecane∣n-Dodecene∣n-Heptane∣n-Heptene∣n-Hexane∣n-Hexene∣n-Nonane∣n-Nonene∣n-Octane∣n-Octene∣n-Pentane∣n-Pentene∣n-Pentylbenzene∣n-Pentylcyclohexane∣n-Undecane∣n-Undecene∣Propylbenzene∣Propylcyclohexane∣p-Xylene (1,4-Dimethylbenzene)∣Toluene
Shipping Condition Room Temperature
Solution Type Mixture
Solvent or Matrix Hydrocarbons
Industry Type Petrochemical Testing
Quantity 10 x 1 mL
Concentration or Composition (by Analyte or Components) n-Decene 1.061 wt%∣Methylcyclohexane 1.248 wt%∣n-Nonene 1.303 wt%∣n-Butylcyclohexane 1.466 wt%∣2,3,3 Trimethyl-1-butene 1.564 wt%∣n-Pentene 1.652 wt%∣Propylcyclohexane 1.711 wt%∣n-Hexene 1.723 wt%∣Decahydronaphthalene 1.768 wt%∣n-Pentylbenzene 1.808 wt%∣n-Pentylcyclohexane 1.876 wt%∣2,3-Dimethyl-1-butene 1.926 wt%∣n-Dodecene 1.927 wt%∣Cyclohexane 2.014 wt%∣n-Pentane 2.176 wt%∣n-Octene 2.193 wt%∣n-Undecene 2.211 wt%∣Isobutylbenzene 2.302 wt%∣Toluene 2.31 wt%∣n-Undecane 2.312 wt%∣Propylbenzene 2.314 wt%∣n-Decane 2.315 wt%∣Benzene 2.318 wt%∣1,2,4,5-Tetramethylbenzene 2.319 wt%∣2,2,4-Trimethylpentane 2.321 wt%∣Ethylbenzene 2.325 wt%∣n-Heptane 2.325 wt%∣Isopropylbenzene 2.33 wt%∣2-Methyl-1-Heptene 2.334 wt%∣2,2-Dimethylbutane 2.335 wt%∣2,4-Dimethylpentane 2.336 wt%∣n-Hexane 2.336 wt%∣2-Methylbutane 2.342 wt%∣1,2,4-Trimethylbenzene 2.345 wt%∣3-Ethyltoluene 2.346 wt%∣n-Butylbenzene 2.348 wt%∣n-Nonane 2.364 wt%∣n-Octane 2.372 wt%∣n-Dodecane 2.377 wt%∣3-Methylpentane 2.39 wt%∣2,3-Dimethylpentane 2.423 wt%∣2-Methylpentane 2.446 wt%∣n-Heptene 2.811 wt%∣Ethylcyclohexane 3.27 wt%∣p-Xylene (1,4-Dimethylbenzene) 3.32 wt%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.