missing translation for 'onlineSavingsMsg'
Learn More

PIANO Isoparaffins Standard

Product Code. 30316452
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30316452 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30316452 Supplier VHG Labs Supplier No. VHGPIANOISO0.1
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,2,3-Trimethylbutane∣2,2,3-Trimethylpentane∣2,2-Dimethylhexane∣2,2-Dimethyloctane∣2,2-Dimethylpentane∣2,3-Dimethylbutane∣2,3-Dimethylheptane∣2,3-Dimethylhexane∣2,3-Dimethyloctane∣2,3-Dimethylpentane∣2,4-Dimethylhexane∣2,4-Dimethylpentane∣2,5-Dimethylheptane∣2,5-Dimethylhexane∣2-Methylbutane∣2-Methylheptane∣2-Methylhexane∣2-Methylnonane∣2-Methyloctane∣2-Methylpentane∣3,3-Diethylpentane∣3,3-Dimethylheptane∣3,3-Dimethyloctane∣3,3-Dimethylpentane∣3,4-Dimethylheptane∣3,5-Dimethylheptane∣3-Ethylhexane∣3-Ethyloctane∣3-Ethylpentane∣3-Methylheptane∣3-Methylhexane∣3-Methylnonane∣3-Methyloctane∣3-Methylpentane∣4-Methylheptane
CAS 464-06-2, 564-02-3, 590-73-8, 15869-87-1, 590-35-2, 79-29-8
Analyte or Component Names 2,2,3-Trimethylbutane∣2,2,3-Trimethylpentane∣2,2-Dimethylhexane∣2,2-Dimethyloctane∣2,2-Dimethylpentane∣2,3-Dimethylbutane∣2,3-Dimethylheptane∣2,3-Dimethylhexane∣2,3-Dimethyloctane∣2,3-Dimethylpentane∣2,4-Dimethylhexane∣2,4-Dimethylpentane∣2,5-Dimethylheptane∣2,5-Dimethylhexane∣2-Methylbutane∣2-Methylheptane∣2-Methylhexane∣2-Methylnonane∣2-Methyloctane∣2-Methylpentane∣3,3-Diethylpentane∣3,3-Dimethylheptane∣3,3-Dimethyloctane∣3,3-Dimethylpentane∣3,4-Dimethylheptane∣3,5-Dimethylheptane∣3-Ethylhexane∣3-Ethyloctane∣3-Ethylpentane∣3-Methylheptane∣3-Methylhexane∣3-Methylnonane∣3-Methyloctane∣3-Methylpentane∣4-Methylheptane
Shipping Condition Room Temperature
Solution Type Mixture
Solvent or Matrix Hydrocarbons
Industry Type Petrochemical Testing
Quantity 0.1 mL
Concentration or Composition (by Analyte or Components) 2,3-Dimethylbutane 0.4421 wt%∣3-Ethylpentane 0.5021 wt%∣3-Ethylhexane 0.6916 wt%∣3,5-Dimethylheptane 0.7506 wt%∣2,2-Dimethylhexane 1.2858 wt%∣2,3-Dimethylheptane 1.4676 wt%∣3,3-Diethylpentane 1.556 wt%∣3-Methylhexane 1.584 wt%∣2,3-Dimethylhexane 1.5959 wt%∣2,4-Dimethylhexane 1.6015 wt%∣3,3-Dimethylheptane 1.6794 wt%∣2,2,3-Trimethylpentane 1.7164 wt%∣2,2-Dimethylpentane 1.7468 wt%∣2,3-Dimethylpentane 1.7627 wt%∣3,3-Dimethylpentane 1.8194 wt%∣2-Methylbutane 2.161 wt%∣2-Methylhexane 2.3496 wt%∣4-Methylheptane 3.1563 wt%∣3,3-Dimethyloctane 3.2087 wt%∣2-Methylpentane 3.236 wt%∣2,2-Dimethyloctane 3.3655 wt%∣2,4-Dimethylpentane 3.6447 wt%∣2,5-Dimethylhexane 3.646 wt%∣2-Methylnonane 3.658 wt%∣3,4-Dimethylheptane 3.6903 wt%∣3-Ethyloctane 3.6912 wt%∣2-Methyloctane 3.7023 wt%∣2,3-Dimethyloctane 3.8263 wt%∣2,2,3-Trimethylbutane 3.8737 wt%∣2-Methylheptane 4.3437 wt%∣3-Methylpentane 5.3104 wt%∣3-Methylheptane 5.4371 wt%∣3-Methyloctane 5.5184 wt%∣2,5-Dimethylheptane 5.5902 wt%∣3-Methylnonane 5.6819 wt%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.