missing translation for 'onlineSavingsMsg'
Learn More

PIANO Olefins Standard

Product Code. 30315848
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30315848 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30315848 Supplier VHG Labs Supplier No. VHGPIANOOLE0.1
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Decene∣1-Heptene∣1-Hexene∣1-Nonene∣1-Octene∣1-Pentene∣2-Methyl-1-butene∣2-Methyl-1-nonene∣2-Methylpentene-2∣3-Methyl-1-butene∣4-Methyl-1-pentene∣cis-2-Heptene∣cis-2-Hexene∣cis-2-Nonene∣cis-2-Octene∣cis-2-Pentene∣cis-3-Heptene∣cis-3-Nonene∣Isoprene∣trans-2-Heptene∣trans-2-Hexene∣trans-2-Nonene∣trans-2-Octene∣trans-2-Pentene∣trans-3-Heptene∣trans-3-Nonene
CAS 872-05-9, 592-76-7, 592-41-6, 124-11-8, 111-66-0, 109-67-1
Analyte or Component Names 1-Decene∣1-Heptene∣1-Hexene∣1-Nonene∣1-Octene∣1-Pentene∣2-Methyl-1-butene∣2-Methyl-1-nonene∣2-Methylpentene-2∣3-Methyl-1-butene∣4-Methyl-1-pentene∣cis-2-Heptene∣cis-2-Hexene∣cis-2-Nonene∣cis-2-Octene∣cis-2-Pentene∣cis-3-Heptene∣cis-3-Nonene∣Isoprene∣trans-2-Heptene∣trans-2-Hexene∣trans-2-Nonene∣trans-2-Octene∣trans-2-Pentene∣trans-3-Heptene∣trans-3-Nonene
Shipping Condition Room Temperature
Solution Type Mixture
Solvent or Matrix Hydrocarbons
Industry Type Petrochemical Testing
Quantity 0.1 mL
Concentration or Composition (by Analyte or Components) 2-Methyl-1-butene 1.4281 wt%∣trans-2-Hexene 1.7157 wt%∣trans-2-Pentene 1.8009 wt%∣trans-2-Octene 1.9312 wt%∣3-Methyl-1-butene 1.9533 wt%∣cis-2-Pentene 1.9716 wt%∣trans-2-Nonene 1.9746 wt%∣trans-3-Nonene 1.9829 wt%∣Isoprene 2.3988 %∣cis-2-Nonene 2.7836 wt%∣2-Methylpentene-2 3.3676 wt%∣4-Methyl-1-pentene 3.4208 wt%∣trans-3-Heptene 3.6329 wt%∣trans-2-Heptene 3.6961 wt%∣cis-2-Hexene 3.8589 wt%∣cis-2-Octene 3.9229 wt%∣cis-3-Nonene 3.9761 wt%∣1-Pentene 4.1176 wt%∣2-Methyl-1-nonene 5.5329 wt%∣cis-2-Heptene 5.7336 wt%∣cis-3-Heptene 5.7577 wt%∣1-Hexene 7.0021 wt%∣1-Heptene 7.5618 wt%∣1-Nonene 7.5881 wt%∣1-Octene 7.6374 wt%∣1-Decene 8.145 wt%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.