missing translation for 'onlineSavingsMsg'
Learn More

PIAS2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16845554 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16845554 150 μL 150µL
16855554 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16845554 Supplier Proteintech Supplier No. 160741AP150UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This gene encodes a protein involved in the regulation of transcription factors involved in MAP kinase signaling. The symbol MIZ1 has also been associated with ZBTB17 which is a different gene located on chromosome 1. Two alternatively spliced transcripts encoding different isoforms have been described.

Specifications

Antigen PIAS2
Applications Western Blot
Classification Polyclonal
Concentration 0.17 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PIAS2
Gene Accession No. O75928, Q6AZ28, Q8C5D8
Gene Alias ARIP3, DAB2 interacting protein, DIP, E3 SUMO protein ligase PIAS2, miz, MIZ1, PIAS NY protein, PIAS2, PIASX, PIASX ALPHA, PIASX BETA, SIZ2, ZMIZ4
Gene Symbols Pias2
Host Species Rabbit
Immunogen PIAS2 Fusion Protein Ag9032
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17344, 83422, 9063
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.