missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This gene encodes a protein involved in the regulation of transcription factors involved in MAP kinase signaling. The symbol MIZ1 has also been associated with ZBTB17 which is a different gene located on chromosome 1. Two alternatively spliced transcripts encoding different isoforms have been described.
Specifications
Specifications
| Antigen | PIAS2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.17 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | PIAS2 |
| Gene Accession No. | O75928, Q6AZ28, Q8C5D8 |
| Gene Alias | ARIP3, DAB2 interacting protein, DIP, E3 SUMO protein ligase PIAS2, miz, MIZ1, PIAS NY protein, PIAS2, PIASX, PIASX ALPHA, PIASX BETA, SIZ2, ZMIZ4 |
| Gene Symbols | Pias2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?