missing translation for 'onlineSavingsMsg'
Learn More

Ovalbumin Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 7 publications

Brand:  Novus Biologicals NB600-922SS

 View more versions of this product

Product Code. 18770984

  • 260.00 € / 0.02mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Ovalbumin Polyclonal specifically detects Ovalbumin in Chicken samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Single Cell Western.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Ovalbumin
Polyclonal
Unconjugated
Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Allergen Gal d 2, CAP 2, CAP2, Cytoplasmic antiproteinase 2, Egg albumin, Gal d II, Hen egg white, PI8, Plakalbumin, Protease inhibitor 8, Serpin B8, serpin peptidase inhibitor clade B member 8, SERPINB14, SERPINB8
Rabbit
42.88 kDa
0.02 mg
Primary
Cross reactivity against Ovalbumin from other sources is unknown.
Chicken, Virus
Antisera
Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot
LYOPH
Western Blot 1:500-1:2000, ELISA 1:5000-1:20000, Immunohistochemistry, Immunoprecipitation, Single Cell Western 1:10
P01012
OVAL
Ovalbumin [Hen Egg White] (Uniprot: P01012)
Multi-step
RUO
396058
For 0.1 mg size: Reconstitute with 10 ul deionized water (or equivalent) to a concentration of 10 mg/mL. The 0.02 mg size is in liquid format.
Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only